MilliporeSigma
  • Intracellular degradation of two structurally different polyhydroxyalkanoic acids accumulated in Pseudomonas putida and Pseudomonas citronellolis from mixtures of octanoic acid and 5-phenylvaleric acid.
Intracellular degradation of two structurally different polyhydroxyalkanoic acids accumulated in Pseudomonas putida and Pseudomonas citronellolis from mixtures of octanoic acid and 5-phenylvaleric acid.
International journal of biological macromolecules (2001-11-24)
D M Chung, M H Choi, J J Song, S C Yoon, I K Kang, N E Huh
ABSTRACT
From a set of mixed carbon sources, 5-phenylvaleric acid (PV) and octanoic acid (OA), polyhydroxyalkanoic acid (PHA) was separately accumulated in the two pseudomonads Pseudomonas putida BM01 and Pseudomonas citronellolis (ATCC 13674) to investigate any structural difference between the two PHA accumulated under a similar culture condition using one-step culture technique. The resulting polymers were isolated by chloroform solvent extraction and characterized by fractional precipitation and differential scanning calorimetry. The solvent fractionation analysis showed that the PHA synthesized by P. putida was separated into two fractions, 3-hydroxy-5-phenylvalerate (3HPV))-rich PHA fraction in the precipitate phase and 3-hydroxyoctanoate (3HO)-rich PHA fraction in the solution phase whereas the PHA produced by P. citronellolis exhibited a rather little compositional separation into the two phases. According to the thermal analysis, the P. putida PHA exhibited two glass transitions indicative of the PHA not being homogeneous whereas the P. citronellolis PHA exhibited only one glass transition. It was found that the structural heterogeneity of the P. putida PHA was caused by a significant difference in the assimilation rate between PV and OA. The structural heterogeneity present in the P. putida PHA was also confirmed by a first order degradation kinetics analysis of the PHA in the cells. The two different first-order degradation rate constants (k(1)), 0.087 and 0.015/h for 3HO- and 3HPV-unit, respectively, were observed in a polymer system over the first 20 h of degradation. In the later degradation period, the disappearance rate of 3HO-unit was calculated to be 0.020 h. The k(1) value of 0.083/h, almost the same as for the 3HO-unit in the P. putida PHA, was obtained for the P(3HO) accumulated in P. putida BM01 grown on OA as the only carbon source. In addition, the k(1) value of 0.015/h for the 3HPV-unit in the P. putida PHA, was also close to 0.019/h for the P(3HPV) homopolymer accumulated in P. putida BM01 grown on PV plus butyric acid. On the contrary, the k(1) values for the P. citronellolis PHA were determined to be 0.035 and 0.029/h for 3HO- and 3HPV-unit, respectively, thus these two relatively close values implying a random copolymer nature of the P. citronellolis PHA. In addition, the faster degradation of P(3HO) than P(3HPV) by the intracellular P. putida PHA depolymerase indicates that the enzyme is more specific against the aliphatic PHA than the aromatic PHA.
Sigma-Aldrich
5-Phenylvaleric acid, 99%