MilliporeSigma
  • Rhodium-catalyzed asymmetric 1,4-addition of organoboron reagents to 5,6-dihydro-2(1H)-pyridinones. Asymmetric synthesis of 4-aryl-2-piperidinones.
Rhodium-catalyzed asymmetric 1,4-addition of organoboron reagents to 5,6-dihydro-2(1H)-pyridinones. Asymmetric synthesis of 4-aryl-2-piperidinones.
The Journal of organic chemistry (2001-10-13)
T Senda, M Ogasawara, T Hayashi
ABSTRACT
Catalytic asymmetric synthesis of 4-aryl-2-piperidinones was realized for the first time by asymmetric 1,4-addition of arylboron reagents to 5,6-dihydro-2(1H)-pyridinones in the presence of a chiral bisphosphine-rhodium catalyst. In the reaction introducing 4-fluorophenyl group, the use of 4-fluorophenylboroxine and 1 equiv (to boron) of water at 40 degrees C gave the highest yield of the arylation product with high enantioselectivity (98% ee). The (R)-4-(4-fluorophenyl)-2-piperidinone obtained here is a key intermediate for the synthesis of (-)-Paroxetine.
Sigma-Aldrich
δ-Valerolactam, 98%