MilliporeSigma
  • Inhalation toxicity studies of the alpha,beta-unsaturated ketones: ethyl vinyl ketone.
Inhalation toxicity studies of the alpha,beta-unsaturated ketones: ethyl vinyl ketone.
Inhalation toxicology (2001-08-11)
D L Morgan, S M Ward, R E Wilson, H C Price, R W O'Connor, J C Seely, M L Cunningham
ABSTRACT
The National Toxicology Program is conducting a chemical class study to investigate the structure-activity relationships for the toxicity of alpha,beta-unsaturated ketones. Ethyl vinyl ketone (EVK) was selected for study because it is a representative straight-chain aliphatic alpha,beta-unsaturated ketone with extensive use and widespread exposure. Short-term inhalation studies of EVK were conducted to provide toxicity data for comparison with the related alpha,beta-unsaturated ketones 2-cyclohexene-1-one (CHX) and methyl vinyl ketone (MVK). These data will be used in designing chronic toxicity and carcinogenicity studies of these ketones. Male and female F344 rats and B6C3F1 mice were exposed to 0, 2, 4, or 8 ppm EVK 6 h/day, 5 days/wk for 13 wk. The nasal cavity was the major target organ of EVK in both rats and mice. Pathologic findings in both the olfactory and respiratory epithelium were observed. Lesions consisted primarily of olfactory epithelial necrosis, atrophy and regeneration, and/or hyperplasia and squamous metaplasia of the respiratory epithelium. Squamous metaplasia of the respiratory epithelium was present in all rats and mice exposed to 4 and 8 ppm EVK, and these lesions were more severe in rats than in mice. Few systemic effects were observed in rats and mice exposed to EVK. A transient decrease in total leukocytes due to decrements in lymphocyte and monocyte populations was present in male rats after exposure to 8 ppm for 3 and 21 days; however, this effect was not present after exposure for 13 wk. There were no chemical-related effects on micronucleus formation in mice, or on sperm motility and vaginal cytology in either species. EVK, like other alpha,beta-unsaturated ketones, is a reactive, direct-acting gaseous irritant with toxicity limited primarily to the upper respiratory tract.
Sigma-Aldrich
1-Penten-3-one, contains 0.1% BHT as stabilizer, 97%