MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Fetal bile acid metabolism: analysis of urinary 3beta-monohydroxy-delta(5) bile acid in preterm infants.
Fetal bile acid metabolism: analysis of urinary 3beta-monohydroxy-delta(5) bile acid in preterm infants.
Biology of the neonate (2001-07-28)
Y Yamato, A Kimura, T Inoue, T Kurosawa, H Kato
ABSTRACT
To elucidate the urinary concentration of total bile acids after birth and the profile of the usual and unusual urinary bile acids, especially 3beta-hydroxy-5-cholen-24-oic acid (Delta(5)-3beta-ol), we measured the concentrations of 13 bile acids in the urine from preterm infants vs. full-term controls by gas chromatography-mass spectrometry. The urinary concentration of total bile acids in early preterm infants below 32 weeks of gestational age significantly exceeded that of the late preterm and full-term infants (p < 0.0005). The major urinary bile acids in early preterm infants were cholic acid, 1beta,3alpha,7alpha,12alpha-tetrahydroxy-5beta-cholan-24-oic acid and Delta(5)-3beta-ol. In conclusion, the high urinary concentrations of total bile acids in preterm infants may be due to an overproduction, or more likely to a low hepatic bile acid clearance. An alternative fetal pathway, the acidic pathway, may be a major route of bile acid biosynthesis in preterm infants.