MilliporeSigma
  • Protective effect of harmaline and harmalol against dopamine- and 6-hydroxydopamine-induced oxidative damage of brain mitochondria and synaptosomes, and viability loss of PC12 cells.
Protective effect of harmaline and harmalol against dopamine- and 6-hydroxydopamine-induced oxidative damage of brain mitochondria and synaptosomes, and viability loss of PC12 cells.
The European journal of neuroscience (2001-06-14)
D H Kim, Y Y Jang, E S Han, C S Lee
ABSTRACT
The present study elucidated the protective effect of beta-carbolines (harmaline, harmalol and harmine) against oxidative damage of brain mitochondria, synaptosomes and PC12 cells induced by either dopamine or 6-hydroxydopamine. Harmaline, harmalol and antioxidant enzymes (superoxide dismutase/SOD and catalase) decreased the alteration of mitochondrial swelling and membrane potential induced by 200 microM dopamine or 100 microM 6-hydroxydopamine. Deprenyl attenuated the dopamine-induced mitochondrial dysfunction but did not reduce the effect of 6-hydroxydopamine. While beta-carbolines inhibited the electron flow in mitochondria, they did not enhance the depressant effect of catecholamines. beta-Carbolines and antioxidant enzymes reversed the depression of synaptosomal Ca2+ uptake induced by 10 microM catecholamines. The compounds inhibited the catecholamine-induced thioredoxin reductase inhibition, thiol oxidation and carbonyl formation in mitochondria and synaptosomes. beta-Carbolines decreased the reactive species-induced deoxyribose degradation. Harmaline and harmalol reduced the catecholamine-induced loss of the transmembrane potential and of cell viability in PC12 cells. beta-Carbolines alone did not show a significant cytotoxic effect on PC12 cells. The results suggest that beta-carbolines may attenuate the dopamine- or 6-hydroxydopamine-induced alteration of brain mitochondrial and synaptosomal functions, and viability loss in PC12 cells, by a scavenging action on reactive oxygen species and inhibition of thiol oxidation.