MilliporeSigma
A role for flavin monooxygenase-like enzymes in auxin biosynthesis.
Science (New York, N.Y.) (2001-02-24)
Y Zhao, S K Christensen, C Fankhauser, J R Cashman, J D Cohen, D Weigel, J Chory
ABSTRACT
Although auxin is known to regulate many processes in plant development and has been studied for over a century, the mechanisms whereby plants produce it have remained elusive. Here we report the characterization of a dominant Arabidopsis mutant, yucca, which contains elevated levels of free auxin. YUCCA encodes a flavin monooxygenase-like enzyme and belongs to a family that includes at least nine other homologous Arabidopsis genes, a subset of which appears to have redundant functions. Results from tryptophan analog feeding experiments and biochemical assays indicate that YUCCA catalyzes hydroxylation of the amino group of tryptamine, a rate-limiting step in tryptophan-dependent auxin biosynthesis.
Sigma-Aldrich
5-Methyl-DL-tryptophan, tryptophan analog