MilliporeSigma
  • A highly sensitive spectrophotometric method with solid-phase extraction for the determination of methylmercury in human hair.
A highly sensitive spectrophotometric method with solid-phase extraction for the determination of methylmercury in human hair.
Journal of analytical toxicology (2000-12-08)
H B Li, F Chen, X R Xu
ABSTRACT
Methylmercury is the most toxic among the mercury species. In order to provide a quick method to screen samples for methylmercury, a highly sensitive spectrophotometric method was established. The method was based on formation of the red complex of methylmercury with thio-Michler's ketone, which can be extracted with n-butanol. The organic layer was determined at a maximum absorption wavelength of 564 nm. The Lambert-Beer law was obeyed for methylmercury from 1.00 x 10(-7) mol/L to 2.00 x 10(-5) mol/L with a correlation coefficient of 0.9992. The detection limit was 3 x 10(-8) mol/L. The method was used to determine methylmercury in human hair. The recovery was from 94% to 102%, and the relative standard deviation was 2.5%. The results agreed with those obtained by gas chromatography with electron capture detection.
Sigma-Aldrich
Michler′s ketone, 98%