MilliporeSigma
  • Diphenyl sulfoxides as selective antagonists of the muscarinic M2 receptor.
Diphenyl sulfoxides as selective antagonists of the muscarinic M2 receptor.
Bioorganic & medicinal chemistry letters (2000-10-31)
J A Kozlowski, D B Lowe, H S Guzik, G Zhou, V B Ruperto, R A Duffy, R McQuade, G Crosby, L A Taylor, W Billard, H Binch, J E Lachowicz
ABSTRACT
Structure activity studies on [4-(phenylsulfonyl)phenyl]methylpiperazine led to the discovery of 4-cyclohexyl-alpha-[4-[[4-methoxyphenyl(S)-sufinyl]phenyl]-1-pi perazineacetonitrile, 1, an M2 selective muscarinic antagonist. Affinity at the cloned human M2 receptor was 2.7 nM; the M1/M2 selectivity is 40-fold.
Sigma-Aldrich
Diphenyl sulfoxide, 96%