MilliporeSigma
Clathration-induced asymmetric transformation of cefadroxil.
Organic letters (2000-08-31)
G J Kemperman, J Zhu, A J Klunder, B Zwanenburg
ABSTRACT
[structure: see text] The cephalosporin antibiotic Cefadroxil can be epimerized at the alpha-carbon of its amino acid side chain using pyridoxal as the mediator. By clathration with 2,7-dihydroxynaphthalene, the desired diastereomer can be selectively withdrawn from the equilibrating mixture of epimers. In this way, an asymmetric transformation of Cefadroxil can be accomplished. This opens the possibility of the production of Cefadroxil starting from racemic p-hydroxyphenylglycine, in contrast to the current industrial synthesis that employs the D-amino acid in enantiopure form.
Sigma-Aldrich
2,7-Dihydroxynaphthalene, 97%