MilliporeSigma
  • Determination of clopidol residues in chicken tissues by high-performance liquid chromatography-mass spectrometry.
Determination of clopidol residues in chicken tissues by high-performance liquid chromatography-mass spectrometry.
Journal of chromatography. A (2000-07-15)
G F Pang, Y Z Cao, C L Fan, J J Zhang, X M Li, C Wang
ABSTRACT
A high-performance liquid chromatographic-mass spectrometric (HPLC-MS) method has been developed for determination of clopidol residues in chicken tissues. Samples are extracted with acetonitrile. The extracts are cleaned up on an alumina column followed by an anion-exchange column. The clopidol is separated on a column (150 cmx4.6 mm) of Intertsil by using acetonitrile-water (20:80) as mobile phase. The clopidol was qualitatively identified by molecule mass and determined quantitatively by selected ion monitoring mode at 190 m/z. The recoveries with RSDs ranged from 91.6+/-10.1 to 97.3+/-5.7 at 0.010 to 10.0 mg/kg by spiking three matrices (chicken muscle, liver, and kidney). The limit of detection was 0.005 mg/kg, and the limit of quantification was 0.010 mg/kg.
Product Description
Supelco
Clopidol, VETRANAL®, analytical standard
Pricing and availability is not currently available.