MilliporeSigma
  • Volume sensitive efflux of taurine in HEK293 cells overexpressing phospholemman.
Volume sensitive efflux of taurine in HEK293 cells overexpressing phospholemman.
Biochimica et biophysica acta (2000-04-20)
M Morales-Mulia, H Pasantes-Morales, J Morán
ABSTRACT
The role of the phospholemman (PLM) on the efflux of taurine and chloride induced by swelling was studied in HEK293 cells overexpressing stable transfected PLM. PLM, a substrate for protein kinases C and A, is a protein that induces an anion current in Xenopus oocytes and forms taurine-selective channels in lipid bilayers. Taurine contributes as an osmolyte to regulatory volume decrease (RVD) and is highly permeable through PLM channels in bilayers. In PLM-overexpressing cells the process of RVD was more rapid and efficient (75%) than in control cells (44%). Also, [(3)H]taurine and (125)I efflux induced by hyposmolarity were markedly increased (30-100%) in two subclones of cells overexpressing PLM. This increased efflux was sensitive to the Cl channel blockers DDF, NPPB and DIDS. Acute treatment of control cells with isoproterenol and norepinephrine induced a significant potentiation (50-60%) of [(3)H]taurine release induced by hyposmolarity. In PLM-overexpressing cells the potentiation by these drugs was higher (100%). Insulin induced also an increase in [(3)H]taurine release, but only in PLM-overexpressing cells (50%). These results indicate that PLM may play a role in the RVD and that its phosphorylation may have a physiological significance during this process. The mechanisms involved in this process could include the activation of PLM itself as channel or the modulation of other preexisting channels.
Sigma-Aldrich
1,9-Dideoxyforskolin from Coleus forskohlii, ≥97%, solid