MilliporeSigma
  • Resolution of overlapped peaks of amino acid derivatives in capillary electrophoresis using multivariate curve resolution based on alternating least squares.
Resolution of overlapped peaks of amino acid derivatives in capillary electrophoresis using multivariate curve resolution based on alternating least squares.
Electrophoresis (2000-03-22)
R M Latorre, J Saurina, S Hernández-Cassou
ABSTRACT
The application of chemometric techniques to the resolution of overlapped peaks in capillary electrophoresis (CE) is described. When a physical separation can not be completely accomplished, chemometrics might still resolve the determination of the analytes mathematically. CE with diode array detection can provide a large amount of data consisting of spectra registered over time. In this study, the capillary electrophoretic separation of 1,2-naphthoquinone-4-sulfonate derivatives of amino acids is studied. Most of the common amino acid derivatives can be separated at 30 kV in a fused-silica capillary by using a 40 mM sodium tetraborate + isopropanol (3:1 v/v) solution as background electrolyte. However, peaks of certain derivatives (Phe, His, Leu and Ile) still overlap. A multivariate curve resolution method based on an alternating least squares optimization procedure is used for the resolution of the overlapped electrophoretic peaks. The method takes advantage of spectral and electrophoretic differences of analytes to recover their pure electrophoretic and spectral profiles. In addition, each analyte in the mixture can be quantified using the corresponding standards.
Sigma-Aldrich
1,2-Naphthoquinone-4-sulfonic acid sodium salt, 97%