MilliporeSigma
  • Regioisomeric distribution of cholesteryl linoleate hydroperoxides and hydroxides in plasma from healthy humans provides evidence for free radical-mediated lipid peroxidation in vivo.
Regioisomeric distribution of cholesteryl linoleate hydroperoxides and hydroxides in plasma from healthy humans provides evidence for free radical-mediated lipid peroxidation in vivo.
Journal of lipid research (2000-01-11)
R Mashima, K Onodera, Y Yamamoto
ABSTRACT
We have previously reported the detection of cholesteryl ester hydroperoxides, consisting mainly of cholesteryl linoleate hydroperoxides (Ch18:2-OOH), at nm levels in plasma from healthy humans (Y. Yamamoto and E. Niki, 1989. Biochem. Biophys. Res. Commun. 165: 988-993). To elucidate their production mechanism in vivo, we examined the distribution of Ch18:2-O(O)H regioisomers in blood plasma from nine healthy young subjects using a sequential method consisting of methanol/hexane extraction in the presence of antioxidant, reductant, and internal standard, solid phase extraction to remove unoxidized cholesteryl linoleate, purification by reversed-phase high-performance liquid chromatography (HPLC), and detection by normal phase HPLC. Furthermore, we confirm that little artifactual oxidation of cholesteryl linoleate occurred during analytical procedures indicated by the absence of oxidation products of cholesteryl 11Z,14Z-eicosadienoate (Ch20:2) when provided as an exogenous substrate to the experimental procedure. We detected nm levels of all free radical-mediated oxidation products, 13ZE-, 13EE-, 9-EZ-, and 9-EE-forms of Ch18:2-O(O)H, in blood plasma, whereas the 13ZE-isomer resulting from enzymatic 15-lipoxygenase oxidation was not evident as a major product. These results indicate that free radical chain oxidation of lipids occurs even in healthy young individuals.
Sigma-Aldrich
Cholesteryl linoleate, ≥98% (HPLC; detection at 205 nm)
Pricing and availability is not currently available.