MilliporeSigma
  • Daphnetin: a novel antimalarial agent with in vitro and in vivo activity.
Daphnetin: a novel antimalarial agent with in vitro and in vivo activity.
The American journal of tropical medicine and hygiene (1992-01-01)
Y Z Yang, A Ranz, H Z Pan, Z N Zhang, X B Lin, S R Meshnick
ABSTRACT
Daphnetin is a dihydroxycoumarin that is being used in China for the treatment of coagulation disorders. It is also a chelator and an antioxidant. In vitro, daphnetin causes a 50% inhibition (IC50) of 3H-hypoxanthine incorporation by Plasmodium falciparum at concentrations between 25 and 40 microM. Several related compounds, such as scopoletin, 2, 3-dihydroxybenzoic acid and 3, 4-dihydroxybenzoic acid show no inhibitory activity. The antimalarial activity of daphnetin is inhibited by the addition of iron. Daphnetin does not appear to be an oxidant drug, since it does not spontaneously generate superoxide in vitro. However, it does alkylate bovine serum albumin when incubated in the presence of iron. In vivo, daphnetin significantly prolongs survival of P. yoelli-infected mice.
Sigma-Aldrich
7,8-Dihydroxycoumarin, ≥97%, powder