MilliporeSigma
  • Proconvulsive effect of vasopressin; mediation by a putative V2 receptor subtype in the central nervous system.
Proconvulsive effect of vasopressin; mediation by a putative V2 receptor subtype in the central nervous system.
Brain research (1997-06-06)
G Croiset, D De Wied
ABSTRACT
Subcutaneously (s.c.) administered [Arg8]vasopressin (AVP) potentiated seizures induced by intracerebroventricular (i.c.v.) injection of 1.95 mg pilocarpine (a muscarinic cholinergic agonist). A bell-shaped relation between dose and effect was found. I.c.v. pretreatment with a V1, V2 or oxytocin receptor antagonist was performed to determine whether and what type of receptor is involved in this proconvulsive effect of vasopressin. For these experiments a higher dose of pilocarpine (2.4 mg i.c.v.) was injected. This caused seizures in a slightly but not significantly higher percentage of the rats. A dose-dependent protective action of the V2 receptor antagonist d(CH2),[D-Ile2,Ile4]AVP (effective doses were 25 and 125 ng) on seizures was found. A reduction was observed in the number of animals that developed tonic-clonic convulsions. Neither the V1 receptor antagonist d(CH2)5[Tyr(Me)2]AVP nor the oxytocin receptor antagonist desGly(NH2)9d(CH2)5[Tyr(Me)2Thr4]OVT possessed anti-convulsive activity. Subsequently the type of receptor was studied in detail with fragments of AVP with either V1 or V2 activity. AVP (with V1 and V2 affinity) (1 and 3 microg s.c.) potentiated pilocarpine (1.95 mg) induced seizures. Vasotocin and oxytocin were without effect. Interestingly neither s.c. nor i.c.v. administration of the selective kidney type vasopressin receptor (V2) agonist dDAVP potentiated pilocarpine induced seizures. Several selective antidiuretic agonists (V2), such as d[Val4]AVP, d[Phe2,Val4,D-Arg8]vasopressin (3 microg), [Val4,D-Arg8]vasopressin (3 microg) and d[Val4,D-Arg8]vasopressin (3 microg) were active. Other selective antidiuretic compounds, such as [Val4]AVP, dAVP, d[Tyr(Me)2]AVP and HO[D-Arg8]vasopressin (3 microg) did not influence seizures. These results demonstrate that a combination of substitution of aminoacid 4 (Gln) by Val and to a lesser extent deamination and the D-arginine form yield an active molecule, which can potentiate pilocarpine induced seizures and suggest the existence of a V2 receptor subtype in the brain.
Sigma-Aldrich
[deamino-Cys1, Val4, D-Arg8]-Vasopressin, ≥95%