MilliporeSigma
  • Enhanced electrocatalytic activity of Pt subnanoclusters on graphene nanosheet surface.
Enhanced electrocatalytic activity of Pt subnanoclusters on graphene nanosheet surface.
Nano letters (2009-05-02)
EunJoo Yoo, Tatsuhiro Okata, Tornoki Akita, Masanori Kohyama, Junji Nakamura, Itaru Honma
ABSTRACT
Graphene nanosheet (GNS) gives rise to an extraordinary modification to the properties of Pt cluster electrocatalysts supported on it. The Pt/GNS electrocatalyst revealed an unusually high activity for methanol oxidation reaction compared to Pt/carbon black catalyst. The Pt/GNS electrocatalyst also revealed quite a different characteristic for CO oxidation among the measured catalyst samples. It is found that Pt particles below 0.5 nm in size are formed on GNS, which would acquire the specific electronic structures of Pt, modifying its catalytic activities.
Sigma-Aldrich
Platinum, nanopowder, 200 nm particle size (SEM), 99.9% (metals basis)
Sigma-Aldrich
Diamminedinitritoplatinum(II) solution, 3.4 wt. % in dilute ammonium hydroxide