- Antagonism at GABAB receptors by saclofen and related sulphonic analogues of baclofen and GABA.
Antagonism at GABAB receptors by saclofen and related sulphonic analogues of baclofen and GABA.
Saclofen (the direct sulphonic analogue of baclofen) is a competitive antagonist of baclofen at GABAB receptors in guinea pig ileum and rat cortical slices (estimated pA2 = 5.3), at least twice as potent as 2-hydroxy-saclofen (pA2 = 5). A series of related sulphonic analogues also antagonised baclofen in the guinea pig ileum, including 2-hydroxy-saclofen amide (pA2 = 3.3), 3-amino-2-hydroxy-2-phenyl-propylsulphonic acid (pA2 = 3.5), 3-amino-2-(benzo-(b)-furan-2-yl)-propylsulphonic acid (pA2 = 4.3), and 3-amino-2-(4-chlorophenyl)-prop-1-enesulphonic acid (pA2 = 2.5-3), but none were more active than saclofen which is the most potent specific GABAB antagonist yet found.