MilliporeSigma
  • Spermine and philanthotoxin potentiate excitatory amino acid responses of Xenopus oocytes injected with rat and chick brain RNA.
Spermine and philanthotoxin potentiate excitatory amino acid responses of Xenopus oocytes injected with rat and chick brain RNA.
Neuroscience letters (1990-07-02)
P Brackley, R Goodnow, K Nakanishi, H L Sudan, P N Usherwood
ABSTRACT
The effects of spermine and a synthetic analogue (PhTX-343) of the polyamine amide toxin, delta-philanthotoxin, on the responses of Xenopus oocytes to application of amino acids were examined using voltage clamp. The oocytes were injected with either total rat brain RNA or chick cerebrum, poly(A+)RNA. The responses to N-methyl-D-aspartate and L-kainate were potentiated by low concentrations (10(-11)-10(-7) M) of PhTX-343 and by 10(-5)-10(-4) M spermine. There was variability between oocytes in terms of their responsiveness to these compounds and recovery from their effects was slow and often incomplete. Prolonged or repeated applications of PhTX-343 and spermine eventually resulted in inhibition. Higher concentrations of these compounds always inhibited the responses to acidic amino acids. Low concentrations of PhTX-343 and spermine also potentiated the responses to nicotine and gamma-aminobutyric acid. These results are discussed in terms of the postulated polyamine binding site on the N-methyl-D-aspartate receptor.