MilliporeSigma
  • Novel endo-alpha-N-acetylgalactosaminidases with broader substrate specificity.
Novel endo-alpha-N-acetylgalactosaminidases with broader substrate specificity.
Glycobiology (2008-07-19)
Dimitris Koutsioulis, David Landry, Ellen P Guthrie
ABSTRACT
In an effort to identify novel endo-alpha-N-acetylgalactosaminidases (endo-alpha-GalNAcases), four potential genes were cloned. Three of the expressed proteins EngEF from Enterococcus faecalis, EngPA from Propionibacterium acnes, and EngCP from Clostridium perfringens were purified and characterized. Their substrate specificity was investigated and compared to the commercially available endo-alpha-GalNAcases from Streptococcus pneumoniae (EngSP) and Alcaligenes sp. (EngAL). All enzymes were incubated with various synthetic substrates, and natural glycoproteins and the released sugars were detected by colorimetric assay and thin layer chromatography analysis. The Core 1 disaccharide Gal beta 1,3GalNAc alpha 1pNP was the most rapidly hydrolyzed substrate by all enzymes tested. EngEF exhibited the highest k(cat) for this substrate. EngEF and EngPA were also able to fully hydrolyze the Core 3 disaccharide GlcNAc beta 1,3GalNAc alpha 1pNP. This is the first report of endo-alpha-GalNAcases EngEF and EngPA acting on Core 3 in addition to Core 1 O-glycans. Interestingly, there were no significant differences in transglycosylation activities when Gal beta 1,3GalNAc alpha 1pNP or GlcNAc beta 1,3GalNAc alpha 1pNP was incubated with various 1-alkanols in the presence of the endo-alpha-GalNAcases tested in this work.
Sigma-Aldrich
O-Glycosidase from Streptococcus pneumoniae, recombinant, expressed in E. coli, buffered aqueous solution