MilliporeSigma
Molecular structure of the human muscle-specific enolase gene (ENO3).
The Biochemical journal (1991-04-15)
M Peshavaria, I N Day
ABSTRACT
The single human gene for muscle-specific enolase was isolated and its structure was characterized, from which the mature mRNA transcript and encoded protein were also deduced. The gene contains 12 exons, spans approx. 6 kb and encodes a protein of 433 residues. The gene structure is similar to that found for the rat neuron-specific enolase gene, and the deduced protein aligns precisely with other enolase sequences, including the sequence of the only published crystallized enolase, yeast eno-1. The 5' boundary of the gene includes a 5' non-coding exon and is characterized by an upstream TATA-like box and CpG-rich region. This region contains potential recognition motifs for general transcriptional regulation involving Sp1, activator protein 1 and 2, CCAAT box transcription factor/nuclear factor I and cyclic AMP, and for muscle-specific transcriptional regulation involving a CC(A + T-rich)6GG box, M-CAT-box CAATCCT and two myocyte-specific enhancer-binding factor 1 boxes.
Sigma-Aldrich
Enolase from baker′s yeast (S. cerevisiae), lyophilized powder, ≥50 units/mg protein
Sigma-Aldrich
Neuron-specific enolase from human brain, ≥95% (SDS-PAGE), buffered aqueous solution