MilliporeSigma
  • Fetal hemoglobin induction by the histone deacetylase inhibitor, scriptaid.
Fetal hemoglobin induction by the histone deacetylase inhibitor, scriptaid.
Cellular and molecular biology (Noisy-le-Grand, France) (2005-09-21)
J Johnson, R Hunter, R McElveen, X H Qian, B S Baliga, B S Pace
ABSTRACT
Many different classes of drugs induce fetal hemoglobin (HbF) including histone deacetylase (HDAC) inhibitors such as butyrate and trichostatin A. Although these agents induce gamma-globin expression in culture many are ineffective in vivo, therefore research efforts continue to identify clinically useful fetal globin inducers. We and others demonstrated a role for p38 mitogen activated protein kinase (MAPK) in gamma-globin promoter activation by HDAC inhibitors. In this study we determined the ability of scriptaid, a novel HDAC inhibitor, to induce gamma-globin expression via p38 MAPK signaling. Scriptaid induced gamma-globin in K562 cells and human erythroid progenitors. Furthermore the p38-selective inhibitor SB203580 completely reversed the ability of scriptaid to induce HbF. To test the potential efficacy of scriptaid in humans, in vivo studies were completed in beta-YAC transgenic mice where the gamma-gene is completely silenced. Scriptaid induced reticulocytosis and human gamma-globin mRNA synthesis. At a concentration of 1 mg/kg/day given by intraperitoneal injections twice weekly we observed a significant 1.8-fold increase in gamma-globin mRNA transcripts. The potential for scriptaid as a treatment option for sickle cell disease will be discussed.
Sigma-Aldrich
Scriptaid, ≥95%, solid