MilliporeSigma
  • Sonic hedgehog signaling pathway is activated in ALK-positive anaplastic large cell lymphoma.
Sonic hedgehog signaling pathway is activated in ALK-positive anaplastic large cell lymphoma.
Cancer research (2009-02-27)
Rajesh R Singh, Jeong Hee Cho-Vega, Yogesh Davuluri, Shuguang Ma, Fatan Kasbidi, Cristiane Milito, Patrick A Lennon, Elias Drakos, L Jeffrey Medeiros, Rajyalakshmi Luthra, Francisco Vega
ABSTRACT
Deregulation of the sonic hedgehog (SHH) signaling pathway has been implicated in several cancers but has not been explored in T-cell lymphomas. Here, we report that the SHH/GLI1 signaling pathway is activated in anaplastic lymphoma kinase (ALK)-positive anaplastic large cell lymphoma (ALCL). We show that SHH, but not its transcriptional effector GLI1, is amplified in ALK+ ALCL tumors and cell lines, and that SHH and GLI1 proteins are highly expressed in ALK+ ALCL tumors and cell lines. We also show that inhibition of SHH/GLI1 signaling with cyclopamine-KAAD, as well as silencing GLI1 gene expression by small interfering (si)RNA, decreased cell viability and clonogenicity of ALK+ ALCL cells. Transfection of wild-type or mutant NPM-ALK into 293T cells showed that only wild-type NPM-ALK increased GLI1 protein levels and activated SHH/GLI1 signaling as shown by increase of CCND2 mRNA levels. Inhibition of ALK tyrosine kinase and phosphatidylinositol 3-kinase (PI3K)/AKT or forced expression of pAKT down-regulated or up-regulated SHH/GLI1 signaling, respectively. Inhibition of GSK3beta in 293T cells also increased protein levels of GLI1. In conclusion, the SHH/GLI1 signaling pathway is activated in ALK+ ALCL. SHH/GLI1 activation is the result of SHH gene amplification and is further mediated by NPM-ALK through activation of PI3K/AKT and stabilization of GLI1 protein. There is a positive synergistic effect between the SHH/GLI1 and PI3K/AKT pathways that contributes to the lymphomagenic effect of NPM-ALK.
Sigma-Aldrich
SANT-1, ≥98% (HPLC), powder
Sigma-Aldrich
JAK3 Inhibitor II, The JAK3 Inhibitor II, also referenced under CAS 211555-04-3, controls the biological activity of JAK3. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.