MilliporeSigma
  • Guggulsterone sensitizes glioblastoma cells to Sonic hedgehog inhibitor SANT-1 induced apoptosis in a Ras/NFκB dependent manner.
Guggulsterone sensitizes glioblastoma cells to Sonic hedgehog inhibitor SANT-1 induced apoptosis in a Ras/NFκB dependent manner.
Cancer letters (2013-04-04)
Deobrat Dixit, Ruchi Ghildiyal, Nikhil Ponnor Anto, Sourav Ghosh, Vivek Sharma, Ellora Sen
ABSTRACT
Since Shh pathway effector, Gli1, is overexpressed in gliomas, we investigated the effect of novel Shh inhibitor SANT-1 on glioma cell viability. Though SANT-1 failed to induce apoptosis, it reduced proliferation of glioma stem-like cells. Apart from canonical Shh cascade, Gli1 is also induced by non-canonical pathways including NFκB. Therefore, a combinatorial strategy with Ras/NFκB inhibitor, Guggulsterone, was employed to enhance effectiveness of SANT-1. Guggulsterone inhibited Ras and NFκB activity and sensitized cells to SANT-1 induced apoptosis via intrinsic apoptotic mechanism. Inhibition of either Ras or NFκB activity was sufficient to sensitize cells to SANT-1. Guggulsterone induced ERK activation also contributed to Caspase-9 activation. Since SANT-1 and Guggulsterone differentially target stem-like and non-stem glioma cells respectively, this combination warrants investigation as an effective anti-glioma therapy.
Sigma-Aldrich
SANT-1, ≥98% (HPLC), powder
Pricing and availability is not currently available.