MilliporeSigma
  • Acute administration of SB-277011A, NGB 2904, or BP 897 inhibits cocaine cue-induced reinstatement of drug-seeking behavior in rats: role of dopamine D3 receptors.
Acute administration of SB-277011A, NGB 2904, or BP 897 inhibits cocaine cue-induced reinstatement of drug-seeking behavior in rats: role of dopamine D3 receptors.
Synapse (New York, N.Y.) (2005-04-29)
Jeremy G Gilbert, Amy Hauck Newman, Eliot L Gardner, Charles R Ashby, Christian A Heidbreder, Arlene C Pak, Xiao-Qing Peng, Zheng-Xiong Xi
ABSTRACT
Recent studies have shown that the novel dopamine (DA) D3 receptor antagonists SB-277011A and NGB 2904 inhibit cocaine- and/or stress-induced reinstatement of drug-seeking behavior. The present study sought to determine if SB-277011A, NGB 2904, or BP-897 (a mixed D3 agonist/antagonist) similarly inhibit cocaine-associated cue-induced reinstatement of drug-seeking behavior. Long-Evans rats were allowed to self-administer cocaine. Each cocaine infusion was paired with discrete conditioned cue-light and tone. Subsequently, drug-seeking (i.e., lever-pressing) behavior was extinguished in the absence of cocaine and cocaine-associated cues. Rats were then tested for cue-induced reinstatement of drug-seeking. We found that cocaine-associated cues evoked robust reinstatement of lever-pressing. Acute intraperitoneal (i.p.) administration of SB-277011A (6, 12, or 24 mg/kg) produced a dose-dependent inhibition of cue-induced reinstatement of drug-seeking behavior by 35, 65, and 85%, respectively, compared to vehicle-treated animals. Acute i.p. administration of NGB 2904 (0.1, 1.0, or 5.0 mg/kg) produced a 45, 30, and 70% inhibition of cue-induced reinstatement, respectively, compared to vehicle-treated animals. Acute i.p. administration of either 0.1 or 1 mg/kg of BP 897 did not produce a significant effect on cue-induced reinstatement, whereas a dose of 3 mg/kg produced a 70% inhibition of cue-induced reinstatement. These findings, combined with previous data, suggest that DA D3 receptor antagonism may underlie the inhibitory effects of SB-277011A and NGB 2904 on cocaine cue-induced reinstatement, while the effects of BP 897 may involve D3 and non-D3 receptor mechanisms.
Sigma-Aldrich
SB-277011-A hydrochloride hydrate, ≥98% (HPLC)
Pricing and availability is not currently available.