Merck
  • Identification of highly concentrated dextrose solution (50% dextrose) extravasation and treatment--a clinical report.
Identification of highly concentrated dextrose solution (50% dextrose) extravasation and treatment--a clinical report.
The American journal of emergency medicine (2013-04-23)
Sarah L Lawson, William Brady, Ahmed Mahmoud
ABSTRACT
Treatment for significant hypoglycemia includes administration of dextrose containing agents, including 50% dextrose (D50%W) intravenously. Significant extravasation of D50%W can lead to complications, including skin and soft tissue injury, loss of limb, or death. The aim of this case report, using an interdisciplinary team approach, explores extravasation protocols as well as literature review, is to provide information about the proper use of hyaluronidase in patients with D50%W extravasations. A 46-year-old African American man presented to the emergency department (ED) after blood glucose level was initially 13 mg/dL. Emergency medical service established a large bore intravenous (IV) line in the right antecubital vein and administered a total of 50 g of D50%W. Upon arrival to the ED, the patient's level of consciousness had significantly improved. After arrival to the ED, the patient started complaining of pain in his right arm, near the site of the IV line insertion. On inspection, the IV site was grossly infiltrated. Hospital protocols for hyperosmolar infiltration were used. Extravasation is a common medical complication of infused medications and needs to be properly identified and treated. The multitude of skills from nursing, medicine, and pharmacy ensures that extravasation is managed appropriately and effectively to ensure safety to patients. Recognition, communication, and awareness of the institutional guidelines on how to treat infiltration and extravasation should be encouraged in all ED and intensive care unit medical personnel who deal with a variety of infusions and IV medications that have serious implications if not treated correctly.
Sigma-Aldrich
Glucose stock solution, 1000 mg/dL
SKU
Pack Size
Availability
Price
Quantity
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
D-(+)-Glucose solution, 45% in H2O, sterile-filtered, BioXtra, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hyaluronidase from bovine testes, Type VIII, lyophilized powder, 300-1,000 U/mg
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hyaluronidase from bovine testes, Type IV-S, powder, suitable for mouse embryo cell culture, 750-3000 units/mg solid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hyaluronidase from bovine testes, Type IV-S, lyophilized powder (essentially salt-free), 750-3000 units/mg solid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hyaluronidase from bovine testes, Type I-S, lyophilized powder, 400-1000 units/mg solid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hyaluronidase from bovine testes, Type VI-S, lyophilized powder, 3,000-15,000 units/mg solid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hyaluronidase from sheep testes, Type V, lyophilized powder, ≥1,500 units/mg solid
SKU
Pack Size
Availability
Price
Quantity
Supelco
D-(+)-Glucose solution, 1 mg/mL in 0.1% benzoic acid, standard for enzymatic assay kits GAGO20, GAHK20, STA20, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
D-(+)-Glucose solution, 100 g/L in H2O, sterile-filtered, BioXtra, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hyaluronidase from sheep testes, Type II, lyophilized powder, ≥300 units/mg solid
SKU
Pack Size
Availability
Price
Quantity
USP
Dextrose, United States Pharmacopeia (USP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
SKU
Pack Size
Availability
Price
Quantity
Supelco
D-(+)-Glucose, analytical standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C