MilliporeSigma
  • ATR-FTIR study of the protonation states of the Glu residue in the multicopper oxidases, CueO and bilirubin oxidase.
ATR-FTIR study of the protonation states of the Glu residue in the multicopper oxidases, CueO and bilirubin oxidase.
FEBS letters (2010-08-24)
Masayo Iwaki, Kunishige Kataoka, Tsutomu Kajino, Ryosuke Sugiyama, Hirotoshi Morishita, Takeshi Sakurai
ABSTRACT
Redox-induced protonation state changes of the Glu residue in the multicopper oxidases, CueO and bilirubin oxidase (BO), were studied by attenuated total reflectance-Fourier transform infrared spectroscopy. By monitoring IR bands of the carboxylic acid C=O stretch in the wild-type and Glu-to-Gln mutant enzymes the Glu506 of CueO (Glu463 of BO) was found to be unprotonated in the oxidised and protonated in the reduced forms. The results provided direct evidence for proton uptake by the Glu, suggesting it plays a key role in the proton donation to the activated oxygen species in the catalytic cycle.
Sigma-Aldrich
Bilirubin Oxidase from Myrothecium verrucaria, lyophilized powder, 15-65 units/mg protein
Pricing and availability is not currently available.