MilliporeSigma
  • Development and characterization of choline and L-glutamate biosensor integrated on silicon microprobes for in-vivo monitoring.
Development and characterization of choline and L-glutamate biosensor integrated on silicon microprobes for in-vivo monitoring.
Conference proceedings : ... Annual International Conference of the IEEE Engineering in Medicine and Biology Society. IEEE Engineering in Medicine and Biology Society. Annual Conference (2007-11-16)
Olivier Frey, Peter van der Wal, Nico de Rooij, Milena Koudelka-Hep
ABSTRACT
A silicon microprobe to measure in real-time choline and L-glutamate in brain tissue is described. The biosensors are prepared by coating integrated platinum thin-film electrodes with an enzymatic membrane by electrochemically aided deposition. This method allows a very local and reproducible immobilization of the enzymes and the achievement of high sensitivities as required for in vivo recordings. Moreover, several closely-spaced electrode sites on the same microprobe can be coated with different enzyme membranes without any cross-contamination.
Sigma-Aldrich
L-Glutamate Oxidase from Streptomyces sp., recombinant, expressed in E. coli, lyophilized powder, ≥5.0 units/mg solid