MilliporeSigma
  • Antibacterial activity of casein-derived peptides isolated from rabbit (Oryctolagus cuniculus) milk.
Antibacterial activity of casein-derived peptides isolated from rabbit (Oryctolagus cuniculus) milk.
The Journal of dairy research (2003-06-13)
Maria Baranyi, Ursula Thomas, Antonio Pellegrini
ABSTRACT
Acid-precipitated rabbit 'whole casein' was digested by trypsin, chymotrypsin, pepsin, and clostripain to screen for possible peptides with antibacterial properties. The peptide fragments were separated by reversed-phase chromatography. The collected fractions were pooled and their antibacterial properties tested against Escherichia coli, Bacillus subtilis and Staphylococcus lentus. Three antibacterial peptide fragments derived from tryptic digestion of rabbit casein were isolated and identified. Their sequences were found as follows: HVEQLLR (residues 50-56 of beta-casein), ILPFIQSLFPFAER (residues 64-77 of beta-casein), and FHLGHLK (residues 19-25 of alpha(s1)-casein). The three peptides were synthesized and found to exert antibacterial effect against gram positive bacteria only. Proteolytic digestion of rabbit casein by chymotrypsin, pepsin and clostripain yielded several peptide fragments with antibacterial activity. Since antibiotic peptides can be released from casein during the digestion of milk proteins, our results suggest a possible antibacterial function of rabbit caseins. It is conceivable that antibacterial peptides can be generated by endopeptidases of the mammalian gastrointestinal tract possibly providing protection for new-born rabbits against aggression of micro-organisms.
Sigma-Aldrich
Clostripain from Clostridium histolyticum, ≥20 units/mg solid
Pricing and availability is not currently available.