MilliporeSigma
  • Purification and characterization of maltose phosphorylase from Bacillus sp. RK-1.
Purification and characterization of maltose phosphorylase from Bacillus sp. RK-1.
Bioscience, biotechnology, and biochemistry (2002-02-06)
Y Inoue, K Ishii, T Tomita, F Fukui
ABSTRACT
Bacillus sp. RK-1 was isolated as a bacterium that produced maltose phosphorylase (MPase) in the culture supernatant. Screening was done from among about 400 isolates that could grow at 55 degrees C in a medium containing maltose as the sole carbon source. The enzyme was purified to an electrophoretically homogeneous state and some properties were investigated. The Mr of the enzyme was estimated to be 170 kDa by gel filtration and 88.5 kDa by SDS-PAGE, suggesting that it consisted of two identical subunits. The enzyme showed optimum activity around pH 6.0-7.0 and the optimum temperature was about 65 degrees C. The enzyme was stable in the range of pH 5.5-8.0 after keeping it at 4 degrees C for 24 h and retained the activity up to about 55 degrees C after keeping it for 15 min. This is the first report about an MPase that could be produced in the culture supernatant. Furthermore, these investigations showed that this MPase is one of the most thermostable ones reported so far.
Sigma-Aldrich
Maltose Phosphorylase from Enterococcus sp., recombinant, expressed in E. coli, lyophilized powder