MilliporeSigma
  • Effects of the endothelin-converting enzyme inhibitor SM-19712 in a mouse model of dextran sodium sulfate-induced colitis.
Effects of the endothelin-converting enzyme inhibitor SM-19712 in a mouse model of dextran sodium sulfate-induced colitis.
Inflammatory bowel diseases (2009-02-10)
Seungjun Lee, Patsy R Carter, Megan N Watts, Jianxiong R Bao, Norman R Harris
ABSTRACT
Ingestion by mice of dextran sodium sulfate (DSS) induces colonic vasoconstriction and inflammation, with some of the effects potentially mediated by the vasoconstrictor endothelin-1 (ET-1). In this study, mice given 5% 40 kD DSS for 5-6 days had elevated colonic immunostaining for ET-1 and platelet endothelial cell adhesion molecule-1 (PECAM-1). Increased ET-1 can induce microvascular constriction; however, the increase in PECAM-1 is consistent with angiogenesis that could decrease flow resistance. Our measurements of intestinal blood flow, via infused microspheres, suggests that these 2 factors may offset each other, with only a nonsignificant tendency for a DSS-induced decrease in flow. Daily administration of the endothelin converting enzyme inhibitor SM-19712 (15 mg/kg) attenuated DSS-induced increases in colonic immunostaining of ET-1 and PECAM-1. SM-19712 attenuated histologic signs of tissue injury and inflammation induced by DSS, and decreased the extent of loose stools and fecal blood. However, the inhibitor did not significantly decrease DSS-induced colon shortening or tissue levels of myeloperoxidase (an indicator of neutrophil infiltration).
Sigma-Aldrich
SM-19712 hydrate, ≥98% (HPLC)