MilliporeSigma
  • Phosphorylation of CREB and mechanical hyperalgesia is reversed by blockade of the cAMP pathway in a time-dependent manner after repeated intramuscular acid injections.
Phosphorylation of CREB and mechanical hyperalgesia is reversed by blockade of the cAMP pathway in a time-dependent manner after repeated intramuscular acid injections.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2003-07-05)
Marie K Hoeger-Bement, Kathleen A Sluka
ABSTRACT
Spinal activation of the cAMP pathway produces mechanical hyperalgesia, sensitizes nociceptive spinal neurons, and phosphorylates the transcription factor cAMP-responsive element binding protein (CREB), which initiates gene transcription. This study examined the role of the cAMP pathway in a model of chronic muscle pain by assessing associated behavioral changes and phosphorylation of CREB. Bilateral mechanical hyperalgesia of the paw was induced by administering two injections of acidic saline, 5 d apart, into the gastrocnemius muscle of male Sprague Dawley rats. Interestingly, the increases in immunoreactivity for CREB and phosphorylated CREB (p-CREB) in the spinal dorsal horn occur 24 hr, but not 1 week, after the second injection of acidic saline compared with pH 7.2 intramuscular injections. Spinal blockade of adenylate cyclase prevents the expected increase in p-CREB that occurs after intramuscular acid injection. The reversal of mechanical hyperalgesia by adenylate cyclase or protein kinase A inhibitors spinally follows a similar pattern with reversal at 24 hr, but not 1 week, compared with the vehicle controls. The p-CREB immunoreactivity in the superficial dorsal horn correlates with the mechanical withdrawal threshold such that increases in p-CREB are associated with decreases in threshold. Therefore, activation of the cAMP pathway in the spinal cord phosphorylates CREB and produces mechanical hyperalgesia associated with intramuscular acid injections. The mechanical hyperalgesia and phosphorylation of CREB depend on early activation of the cAMP pathway during the first 24 hr but are independent of the cAMP pathway by 1 week after intramuscular injection of acid.
Sigma-Aldrich
SQ 22,536, ≥97% (HPLC), powder
Pricing and availability is not currently available.