MilliporeSigma
  • Validation of reference genes for the relative quantification of gene expression in human epicardial adipose tissue.
Validation of reference genes for the relative quantification of gene expression in human epicardial adipose tissue.
PloS one (2012-04-19)
Kanta Chechi, Yves Gelinas, Patrick Mathieu, Yves Deshaies, Denis Richard
ABSTRACT
Relative quantification is a commonly used method for assessing gene expression, however its accuracy and reliability is dependent upon the choice of an optimal endogenous control gene, and such choice cannot be made a priori. There is limited information available on suitable reference genes to be used for studies involving human epicardial adipose tissue. The objective of the current study was to evaluate and identify optimal reference genes for use in the relative quantification of gene expression in human epicardial fat depots of lean, overweight and obese subjects. Some of the commonly used reference genes including 18S, ACTB, RPL27, HPRT, CYCA, GAPDH, RPLPO, POLR2A and B2M were quantified using real-time PCR analysis. The expression stability of these genes was evaluated using Genorm, Normfinder and Bestkeeper algorithms. In addition, the effect of sample size on the validation process was studied by randomly categorizing subjects in two cohorts of n = 2 and n = 33. CYCA, GAPDH and RPL27 were identified as the most stable genes common to all three algorithms and both sample sizes. Their use as reference gene pairs might contribute to the enhanced robustness of relative quantification in the studies involving the human epicardial adipose tissue.
Sigma-Aldrich
SYBR® Green JumpStart Taq ReadyMix, for quantitative PCR, without MgCl2 in buffer