MilliporeSigma
  • Evaluation of whole genome amplification protocols for array and oligonucleotide CGH.
Evaluation of whole genome amplification protocols for array and oligonucleotide CGH.
Diagnostic molecular pathology : the American journal of surgical pathology, part B (2007-11-29)
Adam Hittelman, Shivaranjani Sridharan, Ritu Roy, Jane Fridlyand, Massimo Loda, Colin Collins, Pamela L Paris
ABSTRACT
Genome-based technologies such as genomic arrays and next generation sequencing are poised to make significant contributions to clinical oncology. However, translation of these technologies to the clinic will require that they produce high-quality reproducible data from small archived tumor specimens and biopsies. Herein, we report on a systematic and comprehensive microarray analysis comparing multiple whole genome amplification methods using a variety of difficult clinical specimens, including formalin-fixed and paraffin-embedded tissues. Quantitative analysis and clustering suggest that Sigma's whole genome amplification protocol performed best on all specimens and, moreover, worked well with a formalin-fixed, paraffin-embedded biopsy.
Sigma-Aldrich
GenomePlex® Whole Genome Amplification (WGA) Kit, Kit for whole genome amplification from a variety of DNA sources including FFPE tissue
Pricing and availability is not currently available.