MilliporeSigma
Effect of surfactants on wetting of super-hydrophobic surfaces.
Langmuir : the ACS journal of surfaces and colloids (2004-10-20)
R Mohammadi, J Wassink, A Amirfazli
ABSTRACT
The effect of surfactants on wetting behavior of super-hydrophobic surfaces was investigated. Super-hydrophobic surfaces were prepared of alkylketene dimer (AKD) by casting the AKD melt in a specially designed mold. Time-dependent studies were carried out, using the axisymmetric drop shape analysis method for contact angle measurement of pure water on AKD surfaces. The results show that both advancing and receding contact angles of water on the AKD surfaces increase over time ( approximately 3 days) and reach the values of about 164 and 147 degrees , respectively. The increase of contact angles is due to the development of a prickly structure on the surface (verified by scanning electron microscopy), which is responsible for its super-hydrophobicity. Aqueous solutions of sodium acetate, sodium dodecyl sulfate, hexadecyltrimethylammonium bromide, and n-decanoyl-n-methylglucamine were used to investigate the wetting of AKD surfaces. Advancing and receding contact angles for various concentrations of different surfactant solutions were measured. The contact angle results were compared to those of a number of pure liquids with surface tensions similar to those of surfactant solutions. It was found that although the surface tensions of pure liquids and surfactant solutions at high concentrations are similar, the contact angles are very different. Furthermore, the usual behavior of super-hydrophobic surfaces that turn super-hydrophilic when the intrinsic contact angle of liquid on a smooth surface (of identical material) is below 90 degrees was not observed in the presence of surfactants. The difference in the results for pure liquids and surfactant solutions is explained using an adsorption hypothesis.
Sigma-Aldrich
N-Decanoyl-N-methylglucamine, ≥98% (GC)