- Culture and manipulation of embryonic cells.
Culture and manipulation of embryonic cells.
Methods in cell biology (2012-01-10)
Lois G Edgar, Bob Goldstein
PMID22226523
ABSTRACT
The direct manipulation of embryonic cells is an important tool for addressing key questions in cell and developmental biology. C. elegans is relatively unique among genetic model systems in being amenable to manipulation of embryonic cells. Embryonic cell manipulation has allowed the identification of cell interactions by direct means, and it has been an important technique for dissecting mechanisms by which cell fates are specified, cell divisions are oriented, and morphogenesis is accomplished. Here, we present detailed methods for isolating, manipulating and culturing embryonic cells of C. elegans.
MATERIALS