MilliporeSigma
  • An immune paradox: how can the same chemokine axis regulate both immune tolerance and activation?: CCR6/CCL20: a chemokine axis balancing immunological tolerance and inflammation in autoimmune disease.
An immune paradox: how can the same chemokine axis regulate both immune tolerance and activation?: CCR6/CCL20: a chemokine axis balancing immunological tolerance and inflammation in autoimmune disease.
BioEssays : news and reviews in molecular, cellular and developmental biology (2010-10-19)
Iain Comerford, Mark Bunting, Kevin Fenix, Sarah Haylock-Jacobs, Wendel Litchfield, Yuka Harata-Lee, Michelle Turvey, Julie Brazzatti, Carly Gregor, Phillip Nguyen, Ervin Kara, Shaun R McColl
ABSTRACT
Chemokines (chemotactic cytokines) drive and direct leukocyte traffic. New evidence suggests that the unusual CCR6/CCL20 chemokine receptor/ligand axis provides key homing signals for recently identified cells of the adaptive immune system, recruiting both pro-inflammatory and suppressive T cell subsets. Thus CCR6 and CCL20 have been recently implicated in various human pathologies, particularly in autoimmune disease. These studies have revealed that targeting CCR6/CCL20 can enhance or inhibit autoimmune disease depending on the cellular basis of pathogenesis and the cell subtype most affected through different CCR6/CCL20 manipulations. Here, we discuss the significance of this chemokine receptor/ligand axis in immune and inflammatory functions, consider the potential for targeting CCR6/CCL20 in human autoimmunity and propose that the shared evolutionary origins of pro-inflammatory and regulatory T cells may contribute to the reason why both immune activation and regulation might be controlled through the same chemokine pathway.