MilliporeSigma
  • Evaluation of RNA-knockdown strategies for modulation of cytochrome P450 reductase activity in mouse hepatocytes.
Evaluation of RNA-knockdown strategies for modulation of cytochrome P450 reductase activity in mouse hepatocytes.
Journal of RNAi and gene silencing : an international journal of RNA and gene targeting research (2011-02-26)
Neil Cw Mackenzie, Simon G Lillico, Ken Brown, Charles R Wolf, Christopher Ba Whitelaw
ABSTRACT
Transgenic technologies can provide important animal models for studying drug-metabolizing enzymes. Our overall aim was to generate versatile cell and animal systems that exhibited varying levels of cytochrome P450 oxidoreductase (POR) activity, more accurately modelling the human population for pharmacological and toxicology studies. Towards this goal we evaluated RNA-interference constructs designed for use in vitro and in vivo for reducing POR activity in hepatocytes. This study clearly demonstrates that both POR protein level and reductase activity can be significantly knocked down in Hepa-1 cells in vitro, while highlighting the difficulty in predicting knockdown efficiency in transgenic animals. The high levels of embryonic lethality observed, and inability to produce multi-copy transgenic animals indicates that high levels of shRNA expression may be detrimental to embryonic development.
Sigma-Aldrich
M16 Medium, With sodium bicarbonate and lactic acid, without penicillin and streptomycin, liquid, sterile-filtered