MilliporeSigma
Novel mediators of amyloid precursor protein signaling.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2009-12-18)
Andrzej Swistowski, Qiang Zhang, Mark E Orcholski, Danielle Crippen, Cathy Vitelli, Alexei Kurakin, Dale E Bredesen
ABSTRACT
Multiple recent reports implicate amyloid precursor protein (APP) signaling in the pathogenesis of Alzheimer's disease, but the APP-dependent signaling network involved has not been defined. Here, we report a novel consensus sequence for interaction with the PDZ-1 and PDZ-2 domains of the APP-interacting proteins Mint1, Mint2, and Mint3 (X11alpha, X11beta, and X11gamma), and multiple novel interactors for these proteins, with the finding that transcriptional coactivators are highly represented among these interactors. Furthermore, we show that Mint3 interaction with a set of the transcriptional coactivators leads to nuclear localization and transactivation, whereas interaction of the same set with Mint1 or Mint2 prevents nuclear localization and transactivation. These results define new mediators of the signal transduction network mediated by APP.
Sigma-Aldrich
BCIP®/NBT Liquid Substrate System, ready to use solution