MilliporeSigma
  • Valproate reduces retinal ganglion cell apoptosis in rats after optic nerve crush.
Valproate reduces retinal ganglion cell apoptosis in rats after optic nerve crush.
Neural regeneration research (2022-12-27)
Feng Pan, Dan Hu, Li-Juan Sun, Qian Bai, Yu-Sheng Wang, Xu Hou
ABSTRACT
The retinal ganglion cells of the optic nerve have a limited capacity for self-repair after injury. Valproate is a histone deacetylase inhibitor and multitarget drug, which has been demonstrated to protect retinal neurons. In this study, we established rat models of optic nerve-crush injury and injected valproate into the vitreous cavity immediately after modeling. We evaluated changes in the ultrastructure morphology of the endoplasmic reticulum of retinal ganglion cells over time via transmission electron microscope. Immunohistochemistry and western blot assay revealed that valproate upregulated the expression of the endoplasmic reticulum stress marker glucose-regulated protein 78 and downregulated the expression of transcription factor C/EBP homologous protein, phosphorylated eukaryotic translation initiation factor 2α, and caspase-12 in the endoplasmic reticulum of retinal ganglion cells. These findings suggest that valproate reduces apoptosis of retinal ganglion cells in the rat after optic nerve-crush injury by attenuating phosphorylated eukaryotic translation initiation factor 2α-C/EBP homologous protein signaling and caspase-12 activation during endoplasmic reticulum stress. These findings represent a newly discovered mechanism that regulates how valproate protects neurons.
Sigma-Aldrich
Monoclonal Anti-TUBB antibody produced in mouse, clone 3B3, purified immunoglobulin, buffered aqueous solution