MilliporeSigma
  • Inhalation of ACE2-expressing lung exosomes provides prophylactic protection against SARS-CoV-2.
Inhalation of ACE2-expressing lung exosomes provides prophylactic protection against SARS-CoV-2.
Nature communications (2024-03-13)
Zhenzhen Wang, Shiqi Hu, Kristen D Popowski, Shuo Liu, Dashuai Zhu, Xuan Mei, Junlang Li, Yilan Hu, Phuong-Uyen C Dinh, Xiaojie Wang, Ke Cheng
ABSTRACT
Continued emergence of SARS-CoV-2 variants of concern that are capable of escaping vaccine-induced immunity highlights the urgency of developing new COVID-19 therapeutics. An essential mechanism for SARS-CoV-2 infection begins with the viral spike protein binding to the human ACE2. Consequently, inhibiting this interaction becomes a highly promising therapeutic strategy against COVID-19. Herein, we demonstrate that ACE2-expressing human lung spheroid cells (LSC)-derived exosomes (LSC-Exo) could function as a prophylactic agent to bind and neutralize SARS-CoV-2, protecting the host against SARS-CoV-2 infection. Inhalation of LSC-Exo facilitates its deposition and biodistribution throughout the whole lung in a female mouse model. We show that LSC-Exo blocks the interaction of SARS-CoV-2 with host cells in vitro and in vivo by neutralizing the virus. LSC-Exo treatment protects hamsters from SARS-CoV-2-induced disease and reduced viral loads. Furthermore, LSC-Exo intercepts the entry of multiple SARS-CoV-2 variant pseudoviruses in female mice and shows comparable or equal potency against the wild-type strain, demonstrating that LSC-Exo may act as a broad-spectrum protectant against existing and emerging virus variants.
Sigma-Aldrich
Anti-MxA, clone M143 (CL143), clone CL143, from mouse