MilliporeSigma
  • BTRX-246040 Acts Through the Ventrolateral Periaqueductal Gray to Exert Antidepressant-Relevant Actions in Mice.
BTRX-246040 Acts Through the Ventrolateral Periaqueductal Gray to Exert Antidepressant-Relevant Actions in Mice.
The international journal of neuropsychopharmacology (2023-06-07)
Zhenlong Li, Yuanyuan Xu, Rourou Li, Zhenyu Sheng, Xinya Chen, Xueman Liu, Chau-Shoun Lee, Hsien-Yu Peng, Tzer-Bin Lin, Ming-Chun Hsieh, Cheng-Yuan Lai, Han-Fang Wu, Dylan Chou
ABSTRACT
BTRX-246040, a nociceptin/orphanin FQ peptide receptor antagonist, is being developed for the treatment of depressive patients. However, the underlying mechanism of this potential antidepressant is still largely unclear. Here, we studied the antidepressant-related actions of BTRX-246040 in the ventrolateral periaqueductal gray (vlPAG). The tail suspension test, forced swim test, female urine sniffing test, sucrose preference test, and learned helplessness (LH) combined with pharmacological approaches were employed to examine the antidepressant-like effects and drug effects on LH-induced depressive-like behavior in C57BL/6J mice. Electrophysiological recordings in vlPAG neurons were used to study synaptic activity. Intraperitoneal administration of BTRX-246040 produced antidepressant-like behavioral effects in a dose-dependent manner. Systemic BTRX-246040 (10 mg/kg) resulted in an increased frequency and amplitude of miniature excitatory postsynaptic currents (EPSCs) in the vlPAG. Moreover, slice perfusion of BTRX-246040 directly elevated the frequency and amplitude of miniature EPSCs and enhanced the evoked EPSCs in the vlPAG, which were blocked by pretreatment with the nociceptin/orphanin FQ peptide receptor agonist Ro 64-6198. In addition, intra-vlPAG application of BTRX-246040 produced antidepressant-like behavioral effects in a dose-dependent manner. Moreover, intra-vlPAG pretreatment with 6-cyano-7-nitroquinoxaline-2,3-dione reversed both systemic and local BTRX-246040-mediated antidepressant-like behavioral effects. Furthermore, both systemic and local BTRX-246040 decreased the LH phenotype and reduced LH-induced depressive-like behavior. The results suggested that BTRX-246040 may act through the vlPAG to exert antidepressant-relevant actions. The present study provides new insight into a vlPAG-dependent mechanism underlying the antidepressant-like actions of BTRX-246040.
Sigma-Aldrich
Ro 64-6198, ≥98% (HPLC)