MilliporeSigma
  • Quantitative proteomics assay reveals G protein-coupled receptor kinase 4-induced HepG2 cell growth inhibition.
Quantitative proteomics assay reveals G protein-coupled receptor kinase 4-induced HepG2 cell growth inhibition.
Heliyon (2024-04-19)
Yunxiu Luo, Jing Yang, Yan Wang
ABSTRACT
To investigate the biological effects and putative biological mechanism of G protein-coupled receptor kinase 4 (GRK4) on HepG2 cells. Cell proliferation, cycle, and apoptosis were evaluated by Cell Counting Kit-8 and flow cytometry (FCM) in HepG2 cells infected with either the GRK4-overexpressing lentivirus vector (OE) or the negative control lentivirus vector (NC). The protein profiles and differentially expressed proteins (DEPs) of the OE and NC cells were analyzed and compared using the quantitative proteomics technique, and their function, expression, and probable mechanism were investigated using bioinformatic assays and parallel reaction monitoring (PRM). HepG2 cells that received the OE grew more slowly than those that received the NC. FCM revealed that, when compared to the NC cells, the OE cells had undergone S-phase cycle arrest, and neither the OE nor NC cells underwent apoptosis. Among the 7006 proteins that were identified by quantitative proteomics, 403 DEPs were examined based on the filtering parameters, with the expressions of 135 being downregulated and 268 being upregulated. In addition to being involved in the peroxisome proliferator-activated receptor (PPAR) signaling pathway, the DEPs were implicated in the biological processes of cell proliferation, cycle, and metabolism. PRM verified the expressions of DEPs that were connected to the PPAR pathway. This study shows that GRK4 prevents HepG2 cells from proliferating and causes cell cycle arrest in the S-phase, while the PPAR pathway is involved in the regulation of HepG2 cells via GRK4.
Sigma-Aldrich
Anti-GRK4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution