Merck
  • p38 MAP kinase mediates arsenite-induced apoptosis through FOXO3a activation and induction of Bim transcription.
p38 MAP kinase mediates arsenite-induced apoptosis through FOXO3a activation and induction of Bim transcription.
Apoptosis : an international journal on programmed cell death (2008-05-10)
Beibei Cai, Zhengui Xia
ABSTRACT
Sodium arsenite induces apoptosis in PC12 cells by activating the stress-activated p38 MAP kinase and the pro-apoptotic Bcl-2 family protein Bim(EL). However, the relationship between p38 and Bim(EL) in this apoptosis has not been fully defined. Here, we report that sodium arsenite stimulates the protein expression and promoter activity of Bim(EL) in a p38-dependent manner. Sodium arsenite also caused nuclear translocation of FOXO3a, indicative of FOXO3a activation. Addition of a p38 inhibitor prevented FOXO3a nuclear translocation. RNAi knock down of FOXO3a inhibited Bim promoter activity, Bim(EL) protein expression, and arsenite-induced apoptosis. Our data identify p38 activation of FOXO3a and subsequent induction of Bim(EL) expression as a novel apoptotic mechanism. Together with our previous finding that Bim(EL) is phosphorylated and activated by p38, these results demonstrate that p38 induces apoptosis by regulating Bim(EL) at both the transcriptional and post-translational levels.
Sigma-Aldrich
SB 202190, ≥98% (HPLC)
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ~98% (HPLC)
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ≥95% (HPLC)
Sigma-Aldrich
Sodium (meta)arsenite, ≥90%
Sigma-Aldrich
SB 203580, solid, ≥98% (HPLC)