MilliporeSigma
  • Metabolic dissimilarity determines the establishment of cross-feeding interactions in bacteria.
Metabolic dissimilarity determines the establishment of cross-feeding interactions in bacteria.
Current biology : CB (2021-11-04)
Samir Giri, Leonardo Oña, Silvio Waschina, Shraddha Shitut, Ghada Yousif, Christoph Kaleta, Christian Kost
ABSTRACT
The exchange of metabolites among different bacterial genotypes profoundly impacts the structure and function of microbial communities. However, the factors governing the establishment of these cross-feeding interactions remain poorly understood. While shared physiological features may facilitate interactions among more closely related individuals, a lower relatedness should reduce competition and thus increase the potential for synergistic interactions. Here, we investigate how the relationship between a metabolite donor and recipient affects the propensity of strains to engage in unidirectional cross-feeding interactions. For this, we performed pairwise cocultivation experiments between four auxotrophic recipients and 25 species of potential amino acid donors. Auxotrophic recipients grew in the vast majority of pairs tested (63%), suggesting metabolic cross-feeding interactions are readily established. Strikingly, both the phylogenetic distance between donor and recipient and the dissimilarity of their metabolic networks were positively associated with the growth of auxotrophic recipients. Analyzing the co-growth of species from a gut microbial community in silico also revealed that recipient genotypes benefitted more from interacting with metabolically dissimilar partners, thus corroborating the empirical results. Together, our work identifies the metabolic dissimilarity between bacterial genotypes as a key factor determining the establishment of metabolic cross-feeding interactions in microbial communities.
Product Description
Deep well plate, with pyramid bottom, 48 rectangular wells, pyramid bottom clear polypropylene wells, maximum volume 5 mL, 25 ea of, sterile