MilliporeSigma
  • Mechanical conditioning of human mesenchymal stem cells for enhancing vascular regeneration.
Mechanical conditioning of human mesenchymal stem cells for enhancing vascular regeneration.
STAR protocols (2023-03-01)
Miles W Massidda, ByungGee Im, Jason Lee, Aaron B Baker
ABSTRACT
Human mesenchymal stem cells (hMSCs) are an appealing cell type for therapeutic applications but remain limited by poor efficacy in clinical trials. Here, we describe a conditioning technique that enhances the vascular regenerative properties of hMSCs and increases their expression of endothelial cell and pericyte markers. We also describe an alginate gel encapsulation protocol for delivering the conditioned cells. For complete details on the use and execution of this protocol, please refer to Lee et al. (2021).1.
Sigma-Aldrich
Human Mesenchymal Stem Cells (Bone Marrow), Human mesenchymal stem cells derived from bone marrow that have a capacity for expansion in vitro while maintaining their potential for differentiation to multiple lineages including adipocytes, osteoblasts and chondrocytes.