MilliporeSigma
Generation of human embryonic stem cells.
Current protocols in stem cell biology (2011-03-15)
Teija Peura, Julia Schaft, Biljana Dumevska, Tomas Stojanov
ABSTRACT
This unit describes generation of human embryonic stem cell lines from early human embryos. The focus is on actual handling of embryos and early embryonic outgrowths, omitting steps required for actual generation, freezing, and thawing of embryos, as well as further culture and characterization of newly derived stem cells. Hence, the initial culture of embryos to a blastocyst stage is described, followed by removal of the protective zona pellucida layer, isolation of the inner cell mass (ICM), subsequent plating of ICM or whole embryo and, finally, the first few passages of an early embryonic outgrowth. A few alternative procedures for some steps such as zona removal and inner cell mass isolation are described, to allow procedures to be modified according to circumstances.
Sigma-Aldrich
Anti-Human Serum antibody produced in goat, whole antiserum
Pricing and availability is not currently available.