MilliporeSigma
Inflammation promotes synucleinopathy propagation.
Experimental & molecular medicine (2022-12-07)
Tae-Kyung Kim, Eun-Jin Bae, Byung Chul Jung, Minsun Choi, Soo Jean Shin, Sung Jun Park, Jeong Tae Kim, Min Kyo Jung, Ayse Ulusoy, Mi-Young Song, Jun Sung Lee, He-Jin Lee, Donato A Di Monte, Seung-Jae Lee
ABSTRACT
The clinical progression of neurodegenerative diseases correlates with the spread of proteinopathy in the brain. The current understanding of the mechanism of proteinopathy spread is far from complete. Here, we propose that inflammation is fundamental to proteinopathy spread. A sequence variant of α-synuclein (V40G) was much less capable of fibril formation than wild-type α-synuclein (WT-syn) and, when mixed with WT-syn, interfered with its fibrillation. However, when V40G was injected intracerebrally into mice, it induced aggregate spreading even more effectively than WT-syn. Aggregate spreading was preceded by sustained microgliosis and inflammatory responses, which were more robust with V40G than with WT-syn. Oral administration of an anti-inflammatory agent suppressed aggregate spreading, inflammation, and behavioral deficits in mice. Furthermore, exposure of cells to inflammatory cytokines increased the cell-to-cell propagation of α-synuclein. These results suggest that the inflammatory microenvironment is the major driver of the spread of synucleinopathy in the brain.
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
X-34, ≥90% (HPLC)
Sigma-Aldrich
Curcumin, from Curcuma longa (Turmeric), powder