MilliporeSigma
  • Morphological and immunocytochemical investigations on mast cells in porcine ureter.
Morphological and immunocytochemical investigations on mast cells in porcine ureter.
Anatomia, histologia, embryologia (2005-11-18)
A Vodenicharov, R Leiser, M Gulubova, T Vlaykova
ABSTRACT
Morphological, morphometric, histochemical and immunocytochemical investigations on mast cells, located in the wall of ureter of 8 months aged pigs were performed. Mast cells were found in all three layers of ureteral wall, but their distribution was irregular and the number unequal. It was established that alcian blue (AB)-positive mast cells were significantly more than toluidine blue (TB)-positive mast cells. A statistically significant smaller number of both AB and TB-stained mast cells were observed in the tunica mucosa. The largest number of mast cells was found in the tunica muscularis. In the adventitia, mast cells were higher in number in the main connective tissue than in the connective tissue near the blood vessels. Mast cells stained with TB showed variably expressed gamma-metachromasia, which was best visible in those situated in the lamina propria of the mucosa. The prevailing parts of mast cells, however, were AB-positive after AB-safranin staining. This was mostly found in mast cells of the tunica muscularis and in mast cells of perivascular location in the tunica adventitia. Immunocytochemically, mast cells were found to be positive for histamine and vasoactive intestinal polypeptide in the muscle coat, and to histamine in the adventitia, as well. On the basis of obtained results it was presumed that the mast cells in porcine ureter most probably took part not only in keeping of local homeostasis, but played also an important role of mobility of smooth muscle cells in the middle layer of ureter on one hand, and, on the other, in the adventitial blood vessels.
Sigma-Aldrich
Anti-Histamine antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution