MilliporeSigma
Cultural and demic co-diffusion of Tubo Empire on Tibetan Plateau.
iScience (2022-12-31)
Kongyang Zhu, Panxin Du, Jiyuan Li, Jianlin Zhang, Xiaojun Hu, Hailiang Meng, Liang Chen, Boyan Zhou, Xiaomin Yang, Jianxue Xiong, Edward Allen, Xiaoying Ren, Yi Ding, Yiran Xu, Xin Chang, Yao Yu, Sheng Han, Guanghui Dong, Chuan-Chao Wang, Shaoqing Wen
ABSTRACT
A high point of Tibetan Plateau (TP) civilization, the expansive Tubo Empire (618-842 AD) wielded great influence across ancient western China. However, whether the Tubo expansion was cultural or demic remains unclear due to sparse ancient DNA sampling. Here, we reported ten ancient genomes at 0.017- to 0.867-fold coverages from the Dulan site with typical Tubo archaeological culture dating to 1308-1130 BP. Nine individuals from three different grave types have close relationship with previously reported ancient highlanders from the southwestern Himalayas and modern core-Tibetan populations. A Dulan-related Tubo ancestry contributed overwhelmingly (95%-100%) to the formation of modern Tibetans. A genetic outlier with dominant Eurasian steppe-related ancestry suggesting a potential population movement into the Tubo-controlled regions from Central Asia. Together with archeological evidence from burial styles and customs, our study suggested the impact of the Tubo empire on the northeast edge of the TP involved both cultural and demic diffusion.
Sigma-Aldrich
Proteinase K from Tritirachium album
Pricing and availability is not currently available.