MilliporeSigma
  • Glycogen synthase kinase 3β/CCR6-positive bone marrow cells correlate with disease activity in multicentric Castleman disease-TAFRO.
Glycogen synthase kinase 3β/CCR6-positive bone marrow cells correlate with disease activity in multicentric Castleman disease-TAFRO.
British journal of haematology (2021-12-08)
Nobuya Abe, Michihito Kono, Michihiro Kono, Naoki Ohnishi, Tomoya Sato, Masato Tarumi, Masaru Yoshimura, Taiki Sato, Kohei Karino, Yuka Shimizu, Yuichiro Fujieda, Masaru Kato, Rie Hasebe, Kenji Oku, Masaaki Murakami, Tatsuya Atsumi
ABSTRACT
Multicentric Castleman disease-thrombocytopenia, anasarca, reticulin fibrosis of bone marrow, renal dysfunction and organomegaly (MCD-TAFRO)-is an emergent phenotype characterized by lymphoproliferation, fluid collection, hemocytopenia and multiple organopathy. Although studies have demonstrated an aberrant blood cytokine/chemokine profile referred to as "chemokine storm", the pathogenesis remains unclear. We aimed to identify pathogenic key molecules, potential diagnostic targets and therapeutic markers in MCD-TAFRO using serum cytokine/chemokine profiles. We performed the targeted cytokine/chemokine multiplex analysis in six cases of MCD-TAFRO with remission or non-remission status. We observed significant changes in serum concentrations of CCL2, CCL5, and Chitinase-3-like-1 in the MCD-TAFRO patients with active state compared to inactive state. Ingenuity pathway analysis revealed that glycogen synthase kinase 3 (GSK3) and CCR6, which is expressed in megakaryocytes, were detected as upstream positive regulators for activating MCD-TAFRO status. More GSK3β+ CCR6+ cells like megakaryocytes were detected in the bone marrow of patients with MCD-TAFRO than in those with systemic lupus erythematosus, MCD-not otherwise specified or autoimmune haemophagocytic lymphohistiocytosis. The cellularity of GSK3β+ CCR6+ cells was correlated with disease activity, including thrombocytopenia and anaemia. In conclusion, GSK3β and CCR6 of bone marrow cells were potentially involved in the pathogenesis of MCD-TAFRO and may act as diagnostic targets and therapeutic markers.
Sigma-Aldrich
Anti-GSK3B antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.